Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific General Chemistry Reagents
Click to view available options
Quantity:
150 mL
2 x 150 R2mL
450 mL
5 x 500 R1mL
625 mL
Type:
Alkaline Phosphatase R1 Kit
Alkaline Phosphatase R2 Kit
Alkaline Phosphatase Reagent Kit
Description
HA416-R1 and HA416-R2 are for Hitachi 747.
- Methods: p-Npp, kinetic and for Hitachi 747
- Linearity: 0 to 1000U/L
- Wavelength: 405nm
- Stability: 7 days at room temp. and 14 days at 2 to 8°C
- Reaction Temp.: 3°C
- Expected Values: Adults 35-123 U/L, Children have a higher normal value
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Alkaline Phosphatase Reagent Kit |
Formulation | R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives |
For Use With (Application) | Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only |
Quantity | 150 mL |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction