Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Glucose Dry Powder Reagents

Catalog No. 23666275 Shop All Pointe Scientific Products
Click to view available options
Quantity:
10 x 100mL (3.4 oz.)
1000 mL
5 x 600 mL
Form:
Dry Powder
Liquid
Formulation:
Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1%
Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers
pH:
7.3
7.4

Catalog No. 23-666-275

Supplier: Pointe Scientific HG4213000

Includes

  • Glucose Oxidase >15u/ml; Peroxidase (horseradish) 800U/L; 4-
  • Aminoantipyrine 0.38mM; Phosphate Buffer; pH 7.5±0.1; Sodium p-Hydroxybenzoate 5mM; non-reactive Stabilizers and fillers; Sodium Azide 0.1%

Quantify glucose levels in serum samples

  • Method: Oxidase/Trinder, Endpoint
  • Format: Dry powder
  • Wavelength: 500nm
  • Linearity: 500mg/dL
  • Expected Values: 70 to 105mg/dL
  • Reconstituted stability: 30 days at 2 to 8°C

For the quantitative determination of glucose in serum

Specifications

Certifications/Compliance CE Marked
Type Glucose (Oxidase) Reagent Kit
Formulation Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers
For Use With (Application) Quantitative determination of Glucose in Serum, in-vitro diagnostic use only
pH 7.3
Quantity 5 x 600 mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.