Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Glucose (Hexokinase) Liquid Reagents
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
Description
- Reagent requires Glucose Standard (23-666-277)/Chemistry Calibrator (23-666-189)
Specifications
Specifications
Certifications/Compliance | CE Marked |
Content And Storage | 2°C to 8°C |
Type | Glucose (Hexokinase) Reagent Kit |
Formulation | Glucose reagent: Nicotinamide Adenine Dinucleotide 2mM/L, Adenosine Triphosphate 4mM/L, Magnesium 2mM/L, Hexokinase (Yeast) > 2000U/L, Glucose-6-Phosphate Sehydrogenase (microbial) >4000U/L, Stabilizers and preservatives |
For Use With (Application) | Quantitative measurement of Glucose in Serum |
Quantity | 1 x 120 mL |
Wavelength | 340 nm |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction