Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron/TIBC Reagents

Catalog No. p-318899
Click to view available options
Quantity:
400 Tests
60 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Iron/TIBC Reagent Kit
Formulation Iron Buffer contains: Acetic Acid (dilute) 6%, UIBC Buffer contains: Sodium Azide 0.05%, Iron color contains: Ferrozine 16.7mM, Iron standard contains: Ferrous Chloride
For Use With (Application) Quantitative determination of Iron and total Iron-binding capacity in Serum
Includes 1 x 1L Iron Buffer, 1 x 1L UIBC Buffer, 1 x 60mL Iron Color, 1 x 500mL Iron Standard
Quantity 400 Tests
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.