Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Iron-Total Liquid Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
1 x 120 mL Buffer, 1 x 5 mL Color
1 x 500 mL Buffer, 1 x 15 mL Color
5 x 50mL R1, 1 x 50mL R2
Type:
Iron (Ferrozine) Reagent
Total Iron Reagent Set 1
Total Iron Reagent Set 2
Description
Includes
- Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5); surfactant
- Color Reagent: Ferrozine 16.7mM in hydroxylamine hydrochloride
- Method: Ferrozine, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 560nm
- Linearity: 500ug/dL
- Expected Values: 60 to 150ug/dL
For the quantitative determination of iron in serum
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Total Iron Reagent Set 1 |
Formulation | Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, Iron color R2: 3.6mM Ferrozine in Hydroxylamine Hydrochloride |
For Use With (Application) | Quantitative determination of Iron in Serum |
Quantity | 1 x 120 mL Buffer, 1 x 5 mL Color |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction