Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron-Total Reagents for Hitachi Analyzers

Catalog No. p-2717405 Shop All Pointe Scientific Products
Click to view available options
Quantity:
3 x 500 mL R1
5 x 200 R2mL
Formulation:
Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer pH 4.5 with Surfactant
Iron color R2: Ferrozine 3.6mM in Hydroxylamine Hydrochloride

Includes

  • Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5) with surfactant
  • Color Reagent: Ferrozine 3.6mM in hydroxylamine hydrochloride

Requires

Requires calibrator/standard

Quantify iron levels in serum

  • Method: Ferrozine
  • Format: Liquid (ready to use)
  • Linearity: 500ug/dL
  • Expected Values: 60 to 150ug/dL

Applications:

For the quantitative determination of iron in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Total Iron Reagent Kit
Formulation Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer pH 4.5 with Surfactant
For Use With (Application) Quantitative determination of Iron in Serum
Quantity 3 x 500 mL R1
Form Liquid

For in vitro diagnostics use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.