Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific ISE Diluent
For sodium, potassium and chloride analysis with Hitachi™ and Roch™e systems
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | ISE Diluent |
Formulation | Borate Buffer 0.05% nonreactive ingredient Anti-microbial |
For Use With (Application) | Quantitative determination of Sodium, Potassium and Chloride in Serum |
Form | Liquid |
Includes: Borate buffer with anti-microbial (0.05%)
Catalog Number | Mfr. No. | Quantity | ForUseWithEquipment | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | ForUseWithEquipment | Price | Quantity & Availability | ||||
23-666-328
|
Pointe Scientific
HI790DIL |
6 x 2 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
23-666-326
|
Pointe Scientific
HI790DL04 |
6 x 1 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
23-666-329
|
Pointe Scientific
HI990DIL |
5 x 300 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Method: Ion-Selective Electrode (ISE)
- Format: Liquid
ISE Diluent, ISE Internal Reference and ISE 1N KCl are for the quantitative determination of sodium, potassium, and chloride in serum by ion-selective electrode methods on Roche/Hitachi systems
Specifications
CE Marked | |
Borate Buffer 0.05% nonreactive ingredient Anti-microbial | |
Liquid |
ISE Diluent | |
Quantitative determination of Sodium, Potassium and Chloride in Serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title