Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE Internal Reference Reagents

Catalog No. p-2717471
Click to view available options
Quantity:
5 x 600 mL
6 x 2 L

Includes

Borate buffer: Sodium chloride 3.2mM; Sodium bicarbonate 1.3mM; Potassium phosphate 0.15mM; anti-microbial (0.05%)

Used for sodium, potassium and chloride analysis with Hitachi™ and Roche™ systems

  • Method: Ion-Selective Electrode (ISE)
  • Format: Liquid

ISE Diluent, ISE Internal Reference and ISE 1N KCl are for the quantitative determination of sodium, potassium, and chloride in serum by ion-selective electrode methods on Roche/Hitachi systems

Specifications

Certifications/Compliance CE Marked
Type ISE Internal Reference Solution
Formulation ISE Internal Reference: Borate Buffer- Sodium Chloride 3.2mM/L, Sodium Bicarbonate 1.3mM/L, Potassium Phosphate 0.15mM/L, Anti-microbial Nonreactive Ingredient 0.05%
For Use With (Application) Quantitative determination of Sodium, Potassium and Chloride in Serum
Quantity 6 x 2 L
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.