Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific ISE Solution Reagents for Roche Cobas™ Analyzers
Click to view available options
Quantity:
60 mL
125 mL
250 mL
Formulation:
Ammonium Hydrogen Difluoride 0.6%
Aqueous solution containing: Na+ 100mM/L, K+ 7mM/L, Cl- 92mM/L and a preservative
Aqueous solution containing: Na+ 150mM/L, K+ 3mM/L, Cl 138mM/L and a preservative
KCl aqueous solution 3.5mM/L
Type:
ISE Solution Reagent
ISE Standard 1 Solution Reagent
ISE Standard 2 Solution Reagent
Description
- Method: Ion-selective Electrode (ISE)
- Format: Liquid
- Reference Solution contains: KCl 3.5mM
- Standard Solution 1 contains: Na+ 150mM; K+ 3mM; Cl- 138mM; Preservative
- Standard Solution 2 contains: Na+ 100mM; K+ 7mM; Cl- 92mM; Preservative
- Etcher Solution contains: Ammonium hydrogen difluoride 0.6%
For ISE applications on Roche Cobas Chemistry Systems
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | ISE Standard 2 Solution Reagent |
Formulation | Aqueous solution containing: Na+ 100mM/L, K+ 7mM/L, Cl- 92mM/L and a preservative |
For Use With (Application) | To provide calibration points for the Sodium, Potassium and Chloride electrodes on the ISE module |
Quantity | 60 mL |
Form | Liquid |
For in vitro diagnostic use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction