Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific ISE Standard Reagents
Shop All Pointe Scientific Products
Click to view available options
Formulation:
NA+ 120mM/L, K+ 3mM/L, Cl- 80mM/L
NA+ 160mM/L, K+ 7mM/L, Cl- 120mM/L
Type:
ISE Standard High Reagent Kit
ISE Standard Low Reagent Kit
Description
- Method: Ion-Specific Electrode (ISE)
- Format: Liquid (ready to use)
- Configuration: 10 ampules × 2.0mL
- ISE Standard Low contains: Na+ 120.0mM; K+ 3.0mM; Cl- 80.0mM
- ISE Standard High contains: Na+ 160.0mM; K+ 7.0mM; Cl- 120.0mM
- For the standardization of Sodium, Potassium and Chloride on Ion-Selective Electrodes
- Intended for use with Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | ISE Standard Low Reagent Kit |
| Formulation | NA+ 120mM/L, K+ 3mM/L, Cl- 80mM/L |
| For Use With (Application) | Standardization of Sodium, Potassium and Chloride on ion selective electrodes |
| Quantity | 10 x 2 mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction