Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Lactate Dehydrogenase Liquid Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
1 x 120mL R1, 1 x 30mL R2
1 x 500mL R1, 1 x 125mL R2
3 x 120mL R1, 1 x 90mL R2
Description
Includes
Buffer and Co-Enzyme when combined: NAD 5.8mM; L-Lactate 55mM; Buffer (pH 8.95); Non-reactive stabilizers; Sodium azide 0.1%
- Method: Modified Wacker/Tris, Kinetic
- Format: Liquid (ready to use)
- Wavelength: 340nm
- Linearity: 1000 U/L
- Expected Values: 80 to 285 U/L (Male, 37°C); 103 to 227 U/L (Female, 37°C)
For the in vitro quantitative kinetic determination of lactate dehydrogenase activity in serum
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Lactate Dehydrogenase Reagent Kit |
Formulation | NAD 5.8mM, L-Lactate 55mM, 8.95 Buffer pH, non-reactive Stabilizers and Sodium Azide 0.1% as preservative |
For Use With (Application) | Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum |
Quantity | 1 x 120mL R1, 1 x 30mL R2 |
Wavelength | 340 nm |
Form | Liquid |
For in vitro diagnostic use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction