Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Microprotein Reagents

Catalog No. 23666375 Shop All Pointe Scientific Products
Click to view available options
Quantity:
2 x 120mL Reagent, 1 x 15mL Standard
8 x 50mL Reagent, 1 x 15mL Standard
Formulation:
Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants and preservative, Protein standard solution: Contains Albumin 50mg/dL in Saline with Sodium Azide 0.05% as a preservative
Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants, preservative
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

  • Microprotein Reagent includes: Buffer; Pyrogallol Red 0.067mM; Sodium molybdate stabilizer 0.153mM; surfactants; preservative
  • Protein Standard Solution includes: Ablumin 50mg/dL in saline; Sodium azide 0.05%

Quantify total protein levels in urine

  • Method: Pyragallol Red, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 600nm
  • Linearity: 250mg/dL
  • Expected Values: 28 to 141mg (24 hr.)

For the quantitative determination of total protein in urine

Specifications

Certifications/Compliance CE Marked
Type Microprotein Reagent Kit
Formulation Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants, preservative
For Use With (Application) Quantitative determination of total protein in urine
Model 717, 911, 912, 914
Quantity 8 x 50mL Reagent, 1 x 15mL Standard
Form Liquid

For in vitro diagnostic use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.