Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Phosphorus Reagents for Hitachi™ Analyzers
Shop All Pointe Scientific Products
Click to view available options
Quantity:
5 x 300 R2mL
5 x 600 R1mL
8 x 50mL R1, 4 x 50mL R2
Description
Includes
Reagent 1 and Reagent 2 when combined: Ammonium molybdate >0.48mM; Sulfuric acid <220mM; surfactant
Requires
Calibrator/standard
- Method: Phosphomolybdate, Endpoint
- Format: Liquid (ready to use)
- Linearity: 12mg/dL
- Expected Values: 2.5 to 4.8mg/dL
Applications:
For the quantitative determination of inorganic phosphorus in serum on Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Phosphorus Reagent Kit |
| Formulation | Ammonium Molybdate >0.48mM, Sulfuric Acid <220mM with Surfactant |
| For Use With (Application) | Quantitative determination of inorganic Phosphorus in Serum |
| Quantity | 8 x 50mL R1, 4 x 50mL R2 |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction