Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Protein-Total Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
1000 mL
120 mL
5 x 300 R2mL
500 mL
8 x 50mL (1.7 oz.)
Formulation:
R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients
Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients
pH:
13.15
13.2
Type:
Total Protein Reagent Kit
Total Protein Reagent Set
Description
Includes
Sodium hydroxide 600mM; Copper sulfate 12mM; Sodium potassium tartrate 32mM; Potassium iodide 30mM; Non-reactive ingredients
- Method: Biuret, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 540nm
- Linearity: 1.0 to 15.0g/dL
- Expected Values: 6.2 to 8.5g/dL
For the quantitative determination of total protein concentration in serum
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Total Protein Reagent Set |
Formulation | Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients |
For Use With (Application) | Quantitative determination of total Protein concentration in Serum, in-vitro diagnostic use only |
pH | 13.15 |
Quantity | 120 mL |
Form | Liquid |
For in vitro diagnostic use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction