Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Protein-Total Reagents

Catalog No. 23666390
Click to view available options
Quantity:
1000 mL
120 mL
5 x 300 R2mL
500 mL
8 x 50mL (1.7 oz.)
Formulation:
R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients
Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients
pH:
13.15
13.2
Type:
Total Protein Reagent Kit
Total Protein Reagent Set
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Sodium hydroxide 600mM; Copper sulfate 12mM; Sodium potassium tartrate 32mM; Potassium iodide 30mM; Non-reactive ingredients

Quantify total protein in serum

  • Method: Biuret, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 540nm
  • Linearity: 1.0 to 15.0g/dL
  • Expected Values: 6.2 to 8.5g/dL

For the quantitative determination of total protein concentration in serum

Specifications

Certifications/Compliance CE Marked
Type Total Protein Reagent Set
Formulation Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients
For Use With (Application) Quantitative determination of total Protein concentration in Serum, in-vitro diagnostic use only
pH 13.15
Quantity 1000 mL
Form Liquid

For in vitro diagnostic use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.