Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Triglycerides Liquid Reagents

Catalog No. p-319035
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
Type:
Triglyceride Reagent Kit
Triglyceride Reagent Set
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Triglyceride Reagent Set
For Use With (Application) Quantitative determination of Triglyceride in Serum or Plasma
Quantity 1 x 1 L
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.