Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Triglycerides Reagents for Hitachi™ Analyzers
Shop All Pointe Scientific Products
Click to view available options
Quantity:
50 mL
100 mL
600 mL
Description
Includes
4-Chlorophenol 3.5mM; ATP >0.5mM; Magnesium salt 10 mM; 4-Aminophenazone 0.3mM; Glycerol Kinase (microbial) >250U/L; Glycerol Phosphate Oxidase (microbial) >4500U/L; Peroxidase (horseradish) >2000U/L; Lipase (microbial) >200,000U/L; buffer (pH 7.3 ± 0.1); surfactants; stabilizers; preservatives, including sodium azide (0.01%)
Requires
Requires calibrator/standard
- Method: Enzymatic/GPO Trinder, Endpoint
- Format: Liquid (ready to use)
- Linearity: 1000mg/dL
- Expected Values: 44 to 148mg/dL
For the quantitative determination of triglycerides in serum or plasma on Hitachi analyzers
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Triglyceride Reagent Kit |
For Use With (Application) | Quantitative determination of Triglyceride in Serum or Plasma, in-vitro diagnostic use only |
pH | 7.3 |
Quantity | 50 mL |
Form | Liquid |
For in vitro diagnostic use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction