Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLA2 Antibody (OTI3F2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301899 Supplier Novus Biologicals Supplier No. NBP273514APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLA2 Monoclonal antibody specifically detects POLA2 in Human samples. It is validated for Western Blot

Specifications

Antigen POLA2
Applications Western Blot
Classification Monoclonal
Clone OTI3F2
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias DNA polymerase alpha 70 kDa subunit, DNA polymerase alpha subunit B, FLJ21662, FLJ37250, polymerase (DNA directed), alpha 2 (70kD subunit), polymerase (DNA-directed), alpha (70kD)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-284 of human POLA2(NP_002680) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23649
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.