Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLDIP1 Antibody (OTI3B12), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N273515J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273515J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273515J549 Supplier Novus Biologicals Supplier No. NBP273515JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLDIP1 Monoclonal specifically detects POLDIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen POLDIP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B12
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BACURD1, BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1, BTB/POZ domain-containing protein KCTD13, hBACURD1, PDIP1FKSG86, POLDIP1TNFAIP1-like protein, Polymerase delta-interacting protein 1, potassium channel tetramerisation domain containing 13
Gene Symbols KCTD13
Host Species Mouse
Immunogen Full length recombinant protein of human POLDIP1 (NP_849194) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 253980.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.