Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLG2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB057295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057295 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057295 Supplier Novus Biologicals Supplier No. NBP299015V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLG2 Polyclonal specifically detects POLG2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen POLG2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DNA polymerase gamma accessory 55 kDa subunit, DNA polymerase subunit gamma-2, mitochondrial, EC 2.7.7.7, HP55, Mitochondrial DNA polymerase accessory subunit, mitochondrial DNA polymerase subunit gamma-2, mitochondrial DNA polymerase, accessory subunit, MtPolB, MTPOLBPOLG-BETA, p55, PEOA4, POLB, POLGB, PolG-beta, polymerase (DNA directed), gamma 2, accessory subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human POLG2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.