Learn More
Pollen allergen Phl p 1 Antibody (Clone 25), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Pollen allergen Phl p 1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | Clone 25 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Allergen Phl p I, grass pollen allergen |
| Gene Symbols | PHLPI |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.