Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pollen allergen Phl p 1 Human anti-Plant, Clone: 25, Novus Biologicals™
SDP

Catalog No. NB141994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141994 25 μg
NB141993 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB141994 Supplier Novus Biologicals Supplier No. NBP28127525UG
Add to Cart
Add to Cart

Human Monoclonal Antibody

Pollen allergen Phl p 1 Monoclonal specifically detects Pollen allergen Phl p 1 in Plant samples. It is validated for Western Blot, ELISA, Dot Blot.

Specifications

Antigen Pollen allergen Phl p 1
Applications Western Blot, ELISA, Dot Blot
Classification Monoclonal
Clone 25
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Dot Blot
Formulation PBS
Gene Accession No. P43213
Gene Alias Allergen Phl p I, grass pollen allergen
Host Species Human
Immunogen Recombinant Phl p 1, a grass pollen allergen.
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgE κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.