Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLN Antibody (1G2/2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309704 Supplier Novus Biologicals Supplier No. NBP250253MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLN Monoclonal antibody specifically detects POLN in Mouse samples. It is validated for Western Blot

Specifications

Antigen POLN
Applications Western Blot
Classification Monoclonal
Clone 1G2/2
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DNA polymerase N, DNA polymerase nu, DNA polymerase POL4P, EC 2.7.7.7, POL4P, polymerase (DNA directed) nu
Host Species Mouse
Immunogen Recombinant fragment of mouse POLN protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 353497
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.