Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR1D Antibody, Novus Biologicals™
SDP

Catalog No. NBP219884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219884 Supplier Novus Biologicals Supplier No. NBP219884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR1D Polyclonal specifically detects POLR1D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen POLR1D
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9Y2S0
Gene Alias AC19, DNA-directed RNA polymerase I subunit D, DNA-directed RNA polymerases I and III subunit RPAC2, FLJ20616, hRPA19, MGC9850, POLR1C, polymerase (RNA) I polypeptide D, 16kDa, RPA16RNA polymerase I 16 kDa subunit, RPA9, RPAC2RNA polymerases I and III subunit AC2, RPC16, RPO1-3, TCS2
Gene Symbols POLR1D
Host Species Rabbit
Immunogen Full length human POLR1D Recombinant protein.
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51082
Test Specificity Human. Species reactivity based on sequence identity: Mouse (90%), Bovine (94%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.