Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2D Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB044464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044464 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044464 Supplier Novus Biologicals Supplier No. NBP297517H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR2D Polyclonal specifically detects POLR2D in Human samples. It is validated for Western Blot.

Specifications

Antigen POLR2D
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias DNA-directed RNA polymerase II 16 kDa polypeptide, DNA-directed RNA polymerase II subunit D, DNA-directed RNA polymerase II subunit RPB4, HSRBP4, HSRPB4, polymerase (RNA) II (DNA directed) polypeptide D, RBP4, RNA polymerase II 16 kDa subunit, RNA polymerase II subunit B4, RNA polymerase II subunit hsRBP4, RPB16
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human POLR2D fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5433
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.