Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | POLR2D |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
POLR2D Polyclonal specifically detects POLR2D in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
POLR2D | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5433 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NRETIASVRSLLLQKKLHKFELACLANLCPETAEESKALIPSLEGRFEDEELQQILDDIQTKRSFQY | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
DNA-directed RNA polymerase II 16 kDa polypeptide, DNA-directed RNA polymerase II subunit D, DNA-directed RNA polymerase II subunit RPB4, HSRBP4, HSRPB4, polymerase (RNA) II (DNA directed) polypeptide D, RBP4, RNA polymerase II 16 kDa subunit, RNA polymerase II subunit B4, RNA polymerase II subunit hsRBP4, RPB16 | |
POLR2D | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title