Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | POLR2E |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
POLR2E Polyclonal specifically detects POLR2E in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
POLR2E | |
Polyclonal | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
DNA directed RNA polymerase II 23 kda polypeptide, DNA-directed RNA polymerase II 23 kDa polypeptide, DNA-directed RNA polymerase II subunit E, DNA-directed RNA polymerases I, II, and III subunit RPABC1, hRPB25, hsRPB5, polymerase (RNA) II (DNA directed) polypeptide E (25kD), polymerase (RNA) II (DNA directed) polypeptide E, 25kDa, RPABC1, RPB5, RPB5 homolog, XAP4RNA polymerases I, II, and III subunit ABC1 | |
POLR2E | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5434 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELLINITEHELVPEHVVMTKEEVTELLARYKLRENQLPRIQAGDPVARYFGIKRGQVVKIIRPSETAGRY | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title