Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2E Antibody (OTI3B5), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N273518APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273518APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273518APC7 Supplier Novus Biologicals Supplier No. NBP273518APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLR2E Monoclonal specifically detects POLR2E in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen POLR2E
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B5
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DNA directed RNA polymerase II 23 kda polypeptide, DNA-directed RNA polymerase II 23 kDa polypeptide, DNA-directed RNA polymerase II subunit E, DNA-directed RNA polymerases I, II, and III subunit RPABC1, hRPB25, hsRPB5, polymerase (RNA) II (DNA directed) polypeptide E (25kD), polymerase (RNA) II (DNA directed) polypeptide E, 25kDa, RPABC1, RPB5, RPB5 homolog, XAP4RNA polymerases I, II, and III subunit ABC1
Gene Symbols POLR2E
Host Species Mouse
Immunogen Full length recombinant protein of human POLR2E (NP_002686) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5434.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.