Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | POLR2J |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
POLR2J Polyclonal antibody specifically detects POLR2J in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
POLR2J | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuroscience | |
PBS (pH 7.2), 40% Glycerol | |
DNA-directed RNA polymerase II subunit J-1, DNA-directed RNA polymerase II subunit RPB11-a, hRPB14, MGC71910, POLR2J1RNA polymerase II subunit B11-a, polymerase (RNA) II (DNA directed) polypeptide J (13.3kD), polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa, RNA polymerase II 13.3 kDa subunit, RPB11, RPB11A, RPB11m | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DPQVLFAGYKVPHPLEHKIIIRVQTTPDYSPQEAFTNAITDLISELSLLEERFR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
P52435 | |
5439 | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title