Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | POLR2J |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0925
![]() ![]() |
Proteintech Group Inc
164031AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Three genes (POLR2 J1, POLR2 J2, and POLR2 J3) form a cluster of total length of 214 530 bp in the genetic locus 7q22.1 on the long arm of chromosome 7 (contig NT_007933). The fourth gene (POLR2 J4, 31 040 bp) was localized in the cytogenetic locus 7p13 of the short arm of chromosome 7 (contig NT_007819). An analysis enabled us to refine dissimilar experimental data on the mapping of the hRPB11 subunit gene on chromosome 7. In particular, the presence of three sites of its localization according to data on hybridization with fluorescent-labeled probes (the FISH method) was explained. It was established that, upon the expression of the four human POLR2 J genes, at least 14 types of mature mRNAs encoding somewhat differing hRPB11 isoforms can be synthesized. Eleven of these mRNAs were revealed (as full-length copies or clearly identifiable fragments) in the available databases of expressed sequence tags and cDNAs. The most probable scheme of origination of the multiple genes of the POLR2 J family as a result of three consecutive segmented duplications increasing in size was proposed and substantiated. On the basis of the scheme, some assumptions on the pathways of evolution of separate human genes and the mechanisms of generation of protein diversity in higher eukaryotes were madeSpecifications
POLR2J | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O08740, P52435 | |
20022, 288588, 5439 | |
POLR2J Fusion Protein Ag9520 | |
Primary | |
-20°C | |
POLR2J |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hRPB14, POLR2J, POLR2J1, RPB11, RPB11A, RPB11m | |
Polr2j | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title