Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2J2 Antibody (OTI3G4), PerCP, Novus Biologicals™
SDP

Catalog No. NB273520PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273520PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273520PCP Supplier Novus Biologicals Supplier No. NBP273520PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLR2J2 Monoclonal specifically detects POLR2J2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen POLR2J2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3G4
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DNA directed RNA polymerase II polypeptide J-related, DNA-directed RNA polymerase II subunit RPB11-b1, HRPB11B, MGC105050, MGC54043, polymerase (RNA) II (DNA directed) polypeptide J2, RNA polymerase II subunit B11-b1, RPB11, RPB11b1DNA-directed RNA polymerase II subunit J2
Gene Symbols POLR2J2
Host Species Mouse
Immunogen Full length recombinant protein of human POLR2J2 (NP_116581) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 246721.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.