Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2K Antibody, Novus Biologicals™
SDP

Catalog No. NB176834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176834 Supplier Novus Biologicals Supplier No. NBP313147
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POLR2K Polyclonal antibody specifically detects POLR2K in Human samples. It is validated for Western Blot

Specifications

Antigen POLR2K
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias ABC10-alpha, DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide, DNA-directed RNA polymerase II subunit K, DNA-directed RNA polymerases I, II, and III subunit RPABC4, hRPB7.0, hsRPB10a, polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa, RNA polymerase II 7.0 kDa subunit, RNA polymerases I, II, and III subunit ABC4, RPABC4, RPB10alphapolymerase (RNA) II (DNA directed) polypeptide K (7.0kD), RPB12, RPB7.0
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human POLR2K. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5440
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.