Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2K Antibody, Novus Biologicals™
SDP

Catalog No. NBP155166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155166 100 μL
NBP15516620 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP155166 Supplier Novus Biologicals Supplier No. NBP155166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR2K Polyclonal specifically detects POLR2K in Human samples. It is validated for Western Blot.

Specifications

Antigen POLR2K
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P53803
Gene Alias ABC10-alpha, DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide, DNA-directed RNA polymerase II subunit K, DNA-directed RNA polymerases I, II, and III subunit RPABC4, hRPB7.0, hsRPB10a, polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa, RNA polymerase II 7.0 kDa subunit, RNA polymerases I, II, and III subunit ABC4, RPABC4, RPB10alphapolymerase (RNA) II (DNA directed) polypeptide K (7.0kD), RPB12, RPB7.0
Gene Symbols POLR2K
Host Species Rabbit
Immunogen Synthetic peptides corresponding to POLR2K(polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa) The peptide sequence was selected from the middle region of POLR2K (NP_005025). Peptide sequence DTQKDVQPPKQQPMIYICGECHTENEIKSRDPIRCRECGYRIMYKKRTKR.
Molecular Weight of Antigen 7 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5440
Test Specificity This product is specific to Subunit or Isoform: RPABC4.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.