Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2L Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169912 25 μg
NB169911 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB169912 Supplier Novus Biologicals Supplier No. NBP31741825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR2L Polyclonal antibody specifically detects POLR2L in Human samples. It is validated for Immunofluorescence

Specifications

Antigen POLR2L
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias DNA-directed RNA polymerase III subunit L, DNA-directed RNA polymerases I, II, and III subunit RPABC5, EC 2.7.7.6, hRPB7.6, hsRPB10b, polymerase (RNA) II (DNA directed) polypeptide L (7.6kD), polymerase (RNA) II (DNA directed) polypeptide L, 7.6kDa, RBP10, RNA polymerase II 7.6 kDa subunit, RPABC5, RPB10, RPB10 homolog, RPB10beta, RPB7.6RNA polymerases I, II, and III subunit ABC5
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: DALDALGLKRYCCRRMLLAHVDLIEKLLNYA
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5441
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.