Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR3A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP153051

 View more versions of this product

Catalog No. NBP153051


Only null left
Add to Cart

Description

Description

POLR3A Polyclonal specifically detects POLR3A in Human samples. It is validated for Western Blot.
Specifications

Specifications

POLR3A
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
DNA-directed RNA polymerase III largest subunit, DNA-directed RNA polymerase III subunit A, DNA-directed RNA polymerase III subunit RPC1, EC 2.7.7.6, hRPC155, POL3A, polymerase (RNA) III (DNA directed) polypeptide A, 155kDa, RNA polymerase III 155 kDa subunit, RNA polymerase III subunit C160, RNA polymerase III subunit RPC155-D, RPC1, RPC155RNA polymerase III subunit C1
Rabbit
156 kDa
100 μL
Primary
This product is specific to Subunit or Isoform: RPC1.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
O14802
POLR3A
Synthetic peptides corresponding to POLR3A(polymerase (RNA) III (DNA directed) polypeptide A, 155 kDa) The peptide sequence was selected from the middle region of POLR3A (NP_008986). Peptide sequence AYFGQKDSVCGVSECIIMGIPMNIGTGLFKLLHKADRDPNPPKRPLIFDT.
Affinity purified
RUO
11128
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.