Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | POLR3A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1530520
![]() |
Novus Biologicals
NBP15305120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP153051
![]() |
Novus Biologicals
NBP153051 |
100 μL |
Each for $487.50
|
|
|||||
Description
POLR3A Polyclonal specifically detects POLR3A in Human samples. It is validated for Western Blot.Specifications
POLR3A | |
Polyclonal | |
Rabbit | |
O14802 | |
11128 | |
IgG | |
This product is specific to Subunit or Isoform: RPC1. |
Western Blot | |
Unconjugated | |
RUO | |
DNA-directed RNA polymerase III largest subunit, DNA-directed RNA polymerase III subunit A, DNA-directed RNA polymerase III subunit RPC1, EC 2.7.7.6, hRPC155, POL3A, polymerase (RNA) III (DNA directed) polypeptide A, 155kDa, RNA polymerase III 155 kDa subunit, RNA polymerase III subunit C160, RNA polymerase III subunit RPC155-D, RPC1, RPC155RNA polymerase III subunit C1 | |
POLR3A | |
Primary | |
156 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title