Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR3B Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB352465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352465 Supplier Novus Biologicals Supplier No. NBP306317PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR3B Polyclonal antibody specifically detects POLR3B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen POLR3B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C128, DKFZp686B10117, DNA-directed RNA polymerase III 127.6 kDa polypeptide, DNA-directed RNA polymerase III subunit B, DNA-directed RNA polymerase III subunit RPC2, EC 2.7.7.6, FLJ10388, polymerase (RNA) III (DNA directed) polypeptide B, RNA polymerase III subunit C2, RPC2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human POLR3B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55703
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.