Learn More
Description
Specifications
Specifications
| Antigen | POLR3D |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Formulation | TBS and 0.1% BSA with 0.09% Sodium Azide |
| Gene Accession No. | NP_001713.2 |
| Gene Alias | BN51, BN51 (BHK21) temperature sensitivity complementing, BN51TDNA-directed RNA polymerase III 47 kDa polypeptide, DNA-directed RNA polymerase III subunit D, DNA-directed RNA polymerase III subunit RPC4, polymerase (RNA) III (DNA directed) polypeptide D, 44kDa, Protein BN51, RNA polymerase III 47 kDa subunit, RNA polymerase III subunit C4, RPC4, RPC53 homolog, temperature sensitive complementation, cell cycle specific, tsBN51, TSBN51RPC53 |
| Gene Symbols | POLR3D |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
