Learn More
POLR3D Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | POLR3D |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2494
![]() |
Bethyl Laboratories, Inc
IHC00662T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2493
![]() |
Bethyl Laboratories, Inc
IHC00662 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee
This gene complements a temperature-sensitive mutant isolated from the BHK-21 Syrian hamster cell line. It leads to a block in progression through the G1 phase of the cell cycle at nonpermissive temperatures.Specifications
POLR3D | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HOX1C | |
POLR3D | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P05423 | |
661 | |
Between 175 and 225 | |
Primary | |
4° C | |
POLR3D |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.