Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR3E Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB329616 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329616 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329616 Supplier Novus Biologicals Supplier No. NBP297859APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR3E Polyclonal antibody specifically detects POLR3E in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen POLR3E
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DNA-directed RNA polymerase III 80 kDa polypeptide, DNA-directed RNA polymerase III subunit RPC5, FLJ10509, KIAA1452, polymerase (RNA) III (DNA directed) polypeptide E (80kD), RNA polymerase III 80 kDa subunit RPC5, RNA polymerase III subunit C5, RPC5, SIN
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human POLR3E fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55718
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.