Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR3F Antibody, Novus Biologicals™
SDP

Catalog No. NBP219888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219888 Supplier Novus Biologicals Supplier No. NBP219888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR3F Polyclonal specifically detects POLR3F in Human samples. It is validated for Western Blot.

Specifications

Antigen POLR3F
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9H1D9
Gene Alias DNA-directed RNA polymerase III 39 kDa polypeptide, DNA-directed RNA polymerase III subunit F, DNA-directed RNA polymerase III subunit RPC6, DNA-directed RNA polymerases III 39 kDa polypeptide, polymerase (RNA) III (DNA directed) polypeptide F (39 kDa), polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa, RNA polymerase III 39 kDa subunit, RNA polymerase III C39 subunit, RNA polymerase III subunit C6, RPC39MGC13517, RPC6
Gene Symbols POLR3F
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human POLR3F. The exact sequence is proprietary.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10621
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Rat (100%), Bovine (100%), Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.