Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-DL-alanine, 95%, MP Biomedicals™

Catalog No. icn10267780 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg
250 mg

Catalog No. ICN10267780

Supplier: MP Biomedicals, Inc 0210267780

Chemical Identifiers

CAS 25281-63-4
Molecular Formula C3H7NO2
MDL Number MFCD00166369

Specifications

Color White
Assay Percent Range 95%
Quantity 100 mg
Formula Weight 1000 to 5000
Physical Form Powder
Chemical Name or Material Poly-DL-alanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.