Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Poly(I:C)
GREENER_CHOICE

Catalog No. p-4817312
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

TLR3 agonist

Poly(I:C) is a synthetic double-stranded RNA (dsRNA); endosomal Toll-like receptor 3 (TLR3) agonist. It also binds to the cytoplasmic receptors, retinoic acid-inducible gene I (RIG-I) and melanoma differentiation-associated gene 5 (MDA-5), elicits the secretion of type I interferon (IFN) and pro-inflammatory cytokines. Poly(I:C) is commonly used for immune system activation in vivo. Poly(I:C) also promotes maturation of hPSC-derived cardiomyocytes by modulating early Notch signaling and cardiomyogenic transcriptional programs. Poly(I:C) promotes generation of stable, mature dendritic cells in combination with TLR8 agonist CL 075 . Transfection into NIT-1 β cells induce β-cell apoptosis. Commonly used to stimulate immune system activation in vivo. Also promotes maturation of hPSC-derived cardiomyocytes.

To prevent denaturation, this product should be reconstituted in solutions with physiological salt concentrations.

Chemical Identifiers

CAS 24939-03-5
Molecular Formula C19H27N7O16P2

Specifications

Quantity 10 mg
Purity >98%
Molecular Weight (g/mol) 671.406
Chemical Name or Material Poly(I:C)
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.