Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-Ubiquitin Antibody (HWA4C4) - K63 Linkage, Novus Biologicals™
SDP

Catalog No. NBP236523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236523 0.05 mL
NB151708 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236523 Supplier Novus Biologicals Supplier No. NBP236523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Poly-Ubiquitin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HWA4C4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
Formulation PBS with 0.09% Sodium Azide
Gene Alias CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80
Gene Symbols RPS27A
Host Species Mouse
Immunogen The hybridoma secreting the antibody was generated by fusion of splenocytes from Balb/c mice which had received repeated immunisation with a synthetic peptide encompassing the ubiquitin K63-isopeptide motif.
Purification Method Ion exchange chromatography
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity Antibody specifically recognizes K63-linked polyubiquitin chains, but not any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitin chain or monoubiquitin. The specificity of this K63-linked ubiquitin specific monoclonal antibody was demonstrated using synthetic ubiquitin chains, single isopeptide-linked polyubiquitin chains, and with endogenously ubiquitinylated species in both cell lysates and intact cells.
Target Species Non-species specific
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.