Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly-Ubiquitin Antibody (HWA4C4) - K63 Linkage, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $390.00
Specifications
Antigen | Poly-Ubiquitin |
---|---|
Clone | HWA4C4 |
Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Poly-Ubiquitin | |
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
Unconjugated | |
RUO | |
PBS with 0.09% Sodium Azide | |
6233 | |
IgG2a | |
Ion exchange chromatography |
HWA4C4 | |
Monoclonal | |
Mouse | |
Non-species specific | |
CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80 | |
The hybridoma secreting the antibody was generated by fusion of splenocytes from Balb/c mice which had received repeated immunisation with a synthetic peptide encompassing the ubiquitin K63-isopeptide motif. | |
Primary | |
Antibody specifically recognizes K63-linked polyubiquitin chains, but not any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitin chain or monoubiquitin. The specificity of this K63-linked ubiquitin specific monoclonal antibody was demonstrated using synthetic ubiquitin chains, single isopeptide-linked polyubiquitin chains, and with endogenously ubiquitinylated species in both cell lysates and intact cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title