Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly-Ubiquitin Rabbit anti-Human, Mouse, Rat, Clone: 5Y8J0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $494.50
Specifications
Antigen | Poly-Ubiquitin |
---|---|
Clone | 5Y8J0 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Poly-Ubiquitin | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of K48-linkage Specific Ubiquitin (P0CG47).,, Sequence: MQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGGMQIFVKTLTGKTITLEVEPSDTIE | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5Y8J0 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6233 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title