Learn More
Promega PolyATtract™ mRNA Isolation Systems
Catalog No. PRZ5300 Shop All Promega Corporation Products
Click to view available options
Quantity:
15 Preps
Includes:
mRNA Isolation System only
mRNA Isolation System, stand
Catalog No. PR-Z5300
Supplier: Promega Z5300
Only null left
Add to Cart
Add to Cart
The PolyATtract System isolates messenger RNA directly from total RNA.
  • Isolates Highly Pure mRNA from Total RNA
  • Protocol complete in approximately 45 minutes
  • Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
  • Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
  • Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000μg total RNA
Specifications
Content And Storage 2°C to 10°C; Nuclease-free Water 3 x 25mL, SSC (20X) 3 x 1.4mL, Biotinylated Oligo(dT) probe 1 x 50μL, MagneSphere™ technology Magnetic separation stand (two-position) 1 x 1.5mL, Streptavidin MagneSphere™ paramagnetic particles, 15 x 0.6 mL
For Use With (Application) In-vitro translation, RT-PCR and cDNA synthesis
Includes mRNA Isolation System, stand
Quantity 15 Preps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.