Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polycystin-1/PKD1 Antibody (PKS161f/B7), FITC, Novus Biologicals™
SDP

Catalog No. NBP250247F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250247F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250247F Supplier Novus Biologicals Supplier No. NBP250247F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Polycystin-1/PKD1 Monoclonal specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Polycystin-1/PKD1
Applications Immunohistochemistry
Classification Monoclonal
Clone PKS161f/B7
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1
Gene Symbols PKD1
Host Species Mouse
Immunogen Recombinant protein (233 aa)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5310
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.