Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polycystin 2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB136603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136603 Supplier Novus Biologicals Supplier No. NBP306363AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Polycystin 2 Polyclonal specifically detects Polycystin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Polycystin 2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias APKD2, Autosomal dominant polycystic kidney disease type II protein, MGC138466, MGC138468, Pc-2, PC2, PKD4, polycystic kidney disease 2 (autosomal dominant), Polycystic kidney disease 2 protein, polycystin-2, Polycystwin, R48321, transient receptor potential cation channel, subfamily P, member 2, TRPP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Polycystin 2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5311
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.