Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Catalog No. AA43769AK
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L

CAS: 70131-67-8 | HOSi | 45.09 g/mol

Polydimethylsiloxane is frequently used in a liquid containing 4-5% silica gel, emulsifiers and preservatives. It is employed in applications like contact lenses, medical devices, elastomers, in shampoos, food (antifoaming agent), caulking, lubricants, kinetic sand , and heat-resistant tiles. PDMS is a common surfactant, component of defoamers and as an herbicidal penetran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70131-67-8
Molecular Formula HOSi
Molecular Weight (g/mol) 45.09
MDL Number MFCD01325012
InChI Key XQSFXFQDJCDXDT-UHFFFAOYSA-N
Synonym silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
PubChem CID 6327522
IUPAC Name hydroxysilicon
SMILES O[Si]

Specifications

Physical Form Liquid
Density 0.97 g/mL
Solubility Information Soluble in carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Insoluble in water and ethanol.
Quantity 250 mL
Chemical Name or Material Polydimethylsiloxane, hydroxy terminated, M.W. 4,200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.